Product Details

SNP ID
rs78711915
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:87828169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTAAAGCCTTGACCAATCTAAGCA[C/T]CTGTATGGTAAAGAAGAGATTACTC
Phenotype
MIM: 605912
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MEPE PubMed Links

Gene Details

Gene
MEPE
Gene Name
matrix extracellular phosphoglycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184694.2 Intron NP_001171623.1
NM_001184695.1 Intron NP_001171624.1
NM_001184696.1 Intron NP_001171625.1
NM_001184697.1 Intron NP_001171626.1
NM_001291183.1 Intron NP_001278112.1
NM_020203.3 Intron NP_064588.1
XM_006714278.2 Intron XP_006714341.1

View Full Product Details