Product Details

SNP ID
rs76253497
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:148649394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTGTTGATTTGTTTTGTTTTACT[A/G]TAGATTAGGTTGCCTTTTCTACAAT
Phenotype
MIM: 611000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPC2 PubMed Links

Gene Details

Gene
EPC2
Gene Name
enhancer of polycomb homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015630.3 Intron NP_056445.3
XM_011510941.2 Intron XP_011509243.1
XM_011510943.2 Intron XP_011509245.1

View Full Product Details