Product Details
- SNP ID
-
rs79126103
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:118846679 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCACAGGAGCGAGGCAGCGCCTGG[T/C]GCCCGGGTGCCCAACGGGTGGACAG
- Phenotype
-
MIM: 131290
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EN1
PubMed Links
Gene Details
- Gene
- EN1
- Gene Name
- engrailed homeobox 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001426.3 |
1505 |
Silent Mutation |
|
|
NP_001417.3 |
View Full Product Details