Product Details

SNP ID
rs77502057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:3068038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGACAGGACTTTTCTGTCACAT[A/C]TTCCTTCAGCGGCTGATTTTCAAAT
Phenotype
MIM: 607280 MIM: 147851
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CNTN4 PubMed Links

Gene Details

Gene
CNTN4
Gene Name
contactin 4
There are no transcripts associated with this gene.

Gene
CNTN4-AS1
Gene Name
CNTN4 antisense RNA 1
There are no transcripts associated with this gene.

Gene
IL5RA
Gene Name
interleukin 5 receptor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000564.4 4025 UTR 3 NP_000555.2
NM_001243099.1 4025 UTR 3 NP_001230028.1
NM_175724.2 4025 Intron NP_783851.1
NM_175725.2 4025 Intron NP_783852.1
NM_175726.3 4025 UTR 3 NP_783853.1
NM_175727.2 4025 Intron NP_783854.1
NM_175728.2 4025 Intron NP_783855.1
XM_011533677.2 4025 UTR 3 XP_011531979.1
XM_011533678.2 4025 UTR 3 XP_011531980.1

View Full Product Details