Product Details

SNP ID
rs78532586
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:32819704 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACCCCGAGATAGAAGCGGAATA[A/G]AGGGACGACGTTACCTTCACCAGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105377021 PubMed Links

Gene Details

Gene
LOC105377021
Gene Name
uncharacterized LOC105377021
There are no transcripts associated with this gene.

Gene
TRIM71
Gene Name
tripartite motif containing 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039111.2 Intron NP_001034200.1

View Full Product Details