Product Details
- SNP ID
-
rs74901757
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:51895735 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGGAGAAGTCACATCCTTTGCCG[G/T]CCCTGCCTTCTCATCCCTGACAACC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IQCF1
PubMed Links
Gene Details
- Gene
- IQCF1
- Gene Name
- IQ motif containing F1
View Full Product Details