Product Details

SNP ID
rs77545594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21397396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTATTAGCATGGCAACTCATCA[C/T]TGGGACATGTTTTCAGGCAAAATAA
Phenotype
MIM: 610528
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHD8 PubMed Links

Gene Details

Gene
CHD8
Gene Name
chromodomain helicase DNA binding protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170629.1 Intron NP_001164100.1
NM_020920.3 Intron NP_065971.2
Gene
SNORD8
Gene Name
small nucleolar RNA, C/D box 8
There are no transcripts associated with this gene.

Gene
SNORD9
Gene Name
small nucleolar RNA, C/D box 9
There are no transcripts associated with this gene.

View Full Product Details