Product Details

SNP ID
rs79296443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70299818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGACCAACCAGGCCTTGGGATA[C/T]GAGCAGAATGAACAGAGTCTCTTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC20 PubMed Links

Gene Details

Gene
LRRC20
Gene Name
leucine rich repeat containing 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278211.1 2251 UTR 3 NP_001265140.1
NM_001278212.1 2251 UTR 3 NP_001265141.1
NM_001278213.1 2251 UTR 3 NP_001265142.1
NM_001278214.1 2251 UTR 3 NP_001265143.1
NM_018205.3 2251 UTR 3 NP_060675.1
NM_018239.3 2251 UTR 3 NP_060709.2
NM_207119.2 2251 UTR 3 NP_997002.1
XM_017016374.1 2251 UTR 3 XP_016871863.1
XM_017016375.1 2251 UTR 3 XP_016871864.1

View Full Product Details