Product Details

SNP ID
rs78058438
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:39084299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTCGGAGTGAGCGATCTTACAG[C/G]AGCGAGCGGTCGGGGAGCCTGTCTC
Phenotype
MIM: 608271
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MACF1 PubMed Links

Gene Details

Gene
MACF1
Gene Name
microtubule-actin crosslinking factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012090.5 130 Missense Mutation AGC,AGG S27R NP_036222.3
XM_005270694.1 130 Intron XP_005270751.1
XM_005270695.1 130 Intron XP_005270752.1
XM_005270696.1 130 Intron XP_005270753.1
XM_005270697.1 130 Intron XP_005270754.1
XM_005270698.1 130 Intron XP_005270755.1
XM_006710519.1 130 Intron XP_006710582.1
XM_006710520.2 130 Intron XP_006710583.1
XM_006710521.1 130 Intron XP_006710584.1
XM_006710522.1 130 Intron XP_006710585.1
XM_006710524.2 130 Intron XP_006710587.1
XM_006710526.3 130 Missense Mutation AGC,AGG S27R XP_006710589.1
XM_006710528.3 130 Intron XP_006710591.1
XM_006710529.3 130 Intron XP_006710592.1
XM_006710530.3 130 Intron XP_006710593.1
XM_006710531.3 130 Intron XP_006710594.1
XM_006710532.3 130 Intron XP_006710595.1
XM_006710533.2 130 Intron XP_006710596.1
XM_006710535.3 130 Intron XP_006710598.1
XM_006710536.2 130 Intron XP_006710599.1
XM_006710538.2 130 Intron XP_006710601.1
XM_011541141.2 130 Intron XP_011539443.1
XM_011541142.2 130 Intron XP_011539444.1
XM_017000873.1 130 Intron XP_016856362.1
XM_017000874.1 130 Intron XP_016856363.1
XM_017000875.1 130 Missense Mutation AGC,AGG S27R XP_016856364.1
XM_017000876.1 130 Intron XP_016856365.1
XM_017000877.1 130 Intron XP_016856366.1
XM_017000878.1 130 Intron XP_016856367.1
XM_017000879.1 130 Intron XP_016856368.1
XM_017000880.1 130 Intron XP_016856369.1
XM_017000881.1 130 Missense Mutation AGC,AGG S27R XP_016856370.1
XM_017000882.1 130 Intron XP_016856371.1

View Full Product Details