Product Details
- SNP ID
-
rs113873576
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:189676919 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCAGCTTATTATGATACCAGAAT[C/T]GGGCAAATTCTGATCAATATTGACT
- Phenotype
-
MIM: 609803
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANKAR
PubMed Links
Gene Details
- Gene
- ANKAR
- Gene Name
- ankyrin and armadillo repeat containing
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_144708.3 |
1003 |
Silent Mutation |
ATC,ATT |
I143I |
NP_653309.3 |
XM_011510673.2 |
1003 |
Silent Mutation |
ATC,ATT |
I143I |
XP_011508975.1 |
XM_011510674.1 |
1003 |
Silent Mutation |
ATC,ATT |
I143I |
XP_011508976.1 |
XM_011510675.2 |
1003 |
Silent Mutation |
ATC,ATT |
I143I |
XP_011508977.1 |
XM_011510676.2 |
1003 |
Silent Mutation |
ATC,ATT |
I143I |
XP_011508978.1 |
XM_011510677.2 |
1003 |
Silent Mutation |
ATC,ATT |
I143I |
XP_011508979.1 |
XM_011510679.2 |
1003 |
Silent Mutation |
ATC,ATT |
I143I |
XP_011508981.1 |
XM_011510680.1 |
1003 |
Silent Mutation |
ATC,ATT |
I143I |
XP_011508982.1 |
XM_011510681.1 |
1003 |
Silent Mutation |
ATC,ATT |
I143I |
XP_011508983.1 |
XM_011510682.2 |
1003 |
Silent Mutation |
ATC,ATT |
I143I |
XP_011508984.1 |
XM_011510685.2 |
1003 |
UTR 5 |
|
|
XP_011508987.1 |
XM_011510686.2 |
1003 |
Silent Mutation |
ATC,ATT |
I143I |
XP_011508988.1 |
XM_011510687.2 |
1003 |
Intron |
|
|
XP_011508989.1 |
XM_011510688.1 |
1003 |
Silent Mutation |
ATC,ATT |
I143I |
XP_011508990.1 |
XM_011510689.2 |
1003 |
Silent Mutation |
ATC,ATT |
I143I |
XP_011508991.1 |
XM_011510691.2 |
1003 |
Intron |
|
|
XP_011508993.1 |
XM_011510692.2 |
1003 |
Intron |
|
|
XP_011508994.1 |
XM_011510693.2 |
1003 |
Intron |
|
|
XP_011508995.1 |
XM_011510694.2 |
1003 |
Intron |
|
|
XP_011508996.1 |
XM_017003413.1 |
1003 |
Silent Mutation |
ATC,ATT |
I143I |
XP_016858902.1 |
XM_017003414.1 |
1003 |
Silent Mutation |
ATC,ATT |
I143I |
XP_016858903.1 |
XM_017003415.1 |
1003 |
Intron |
|
|
XP_016858904.1 |
XM_017003416.1 |
1003 |
UTR 5 |
|
|
XP_016858905.1 |
XM_017003417.1 |
1003 |
Silent Mutation |
ATC,ATT |
I143I |
XP_016858906.1 |
XM_017003418.1 |
1003 |
Intron |
|
|
XP_016858907.1 |
XM_017003419.1 |
1003 |
Intron |
|
|
XP_016858908.1 |
XM_017003420.1 |
1003 |
Intron |
|
|
XP_016858909.1 |
- Gene
- ASNSD1
- Gene Name
- asparagine synthetase domain containing 1
There are no transcripts associated with this gene.
View Full Product Details