Product Details

SNP ID
rs116482786
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:93934372 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAATTGAAAAAATGTGAAGGTTG[C/T]CAGGATTAAGGAAAACTCAAAGATG
Phenotype
MIM: 176880
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PROS1 PubMed Links

Gene Details

Gene
PROS1
Gene Name
protein S (alpha)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000313.3 Intron NP_000304.2
NM_001314077.1 Intron NP_001301006.1

View Full Product Details