Product Details

SNP ID
rs115365859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:120167084 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAAGTTCACAATAGGCTTGGAG[C/T]GGGCCCACAGCTTTTCCCAGGAAGC
Phenotype
MIM: 610464
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR156 PubMed Links

Gene Details

Gene
GPR156
Gene Name
G protein-coupled receptor 156
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168271.1 2855 Missense Mutation NP_001161743.1
NM_153002.2 2855 Missense Mutation NP_694547.2
XM_005247165.2 2855 Missense Mutation XP_005247222.1
XM_011512484.2 2855 Missense Mutation XP_011510786.1
XM_011512487.1 2855 Missense Mutation XP_011510789.1
XM_011512488.2 2855 Missense Mutation XP_011510790.1
XM_017005795.1 2855 Missense Mutation XP_016861284.1
XM_017005796.1 2855 Missense Mutation XP_016861285.1
XM_017005797.1 2855 Missense Mutation XP_016861286.1

View Full Product Details