Product Details

SNP ID
rs114289133
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:38243518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCATTTTTTTGTTTAATTCCCCA[A/G]TAACACTTAAGTGAGCATTATTACG
Phenotype
MIM: 609642 MIM: 615450
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TARP PubMed Links

Gene Details

Gene
TARP
Gene Name
TCR gamma alternate reading frame protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003799.1 Intron NP_001003799.1
NM_001003806.1 Intron NP_001003806.1
XM_011515407.2 Intron XP_011513709.1
XM_017012226.1 Intron XP_016867715.1
XM_017012227.1 Intron XP_016867716.1
XM_017012228.1 Intron XP_016867717.1
XM_017012229.1 Intron XP_016867718.1
XM_017012230.1 Intron XP_016867719.1
Gene
TRGC2
Gene Name
T cell receptor gamma constant 2
There are no transcripts associated with this gene.

View Full Product Details