Product Details

SNP ID
rs112786144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:83961075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGGCAAATGACATTTTTATTTCT[C/T]AGGCAAAGAAGAAAGACAAACCTGT
Phenotype
MIM: 603961
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEMA3A PubMed Links

Gene Details

Gene
SEMA3A
Gene Name
semaphorin 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006080.2 2962 UTR 3 NP_006071.1
XM_005250110.3 2962 UTR 3 XP_005250167.1
XM_005250111.4 2962 UTR 3 XP_005250168.1
XM_006715839.3 2962 UTR 3 XP_006715902.1
XM_011515734.2 2962 UTR 3 XP_011514036.1
XM_017011673.1 2962 UTR 3 XP_016867162.1

View Full Product Details