Product Details

SNP ID
rs118103386
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:117486989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGTAGTTGAAGGTGAAGGAAAGA[A/G]AAGAGTTAAGGATAACATCTATATT
Phenotype
MIM: 602421
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CFTR PubMed Links
Additional Information
For this assay, SNP(s) [rs138257490] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CFTR
Gene Name
cystic fibrosis transmembrane conductance regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000492.3 Intron NP_000483.3
XM_011515751.2 Intron XP_011514053.1
XM_011515753.1 Intron XP_011514055.1
XM_011515754.2 Intron XP_011514056.1
XM_017011699.1 Intron XP_016867188.1

View Full Product Details