Product Details

SNP ID
rs113378963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:120951247 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCAGGTGCAGAGTAAGTCGGCG[A/C]GTCTACTACTCCTGTTCGCTGCGCG
Phenotype
MIM: 607493
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ING3 PubMed Links

Gene Details

Gene
ING3
Gene Name
inhibitor of growth family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019071.2 Intron NP_061944.2
NM_198267.1 Intron NP_938008.1
XM_017012369.1 Intron XP_016867858.1

View Full Product Details