Product Details

SNP ID
rs117549065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35607904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGAAAACTGTTAATTCTTCCCC[A/G]ACACTATTATCTCTGACCCCCAGGC
Phenotype
MIM: 107272 MIM: 601782
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD72 PubMed Links

Gene Details

Gene
CD72
Gene Name
CD72 molecule
There are no transcripts associated with this gene.

Gene
MIR4667
Gene Name
microRNA 4667
There are no transcripts associated with this gene.

Gene
TESK1
Gene Name
testis-specific kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318230.1 Intron NP_001305159.1
NM_006285.2 Intron NP_006276.2

View Full Product Details