Product Details

SNP ID
rs115131816
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:45772257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAAGGTGGAGGGACCTTGCCTC[C/T]GAGGATTCATCCCACCCGGATTCCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DKFZp779M0652 PubMed Links

Gene Details

Gene
DKFZp779M0652
Gene Name
uncharacterized DKFZp779M0652
There are no transcripts associated with this gene.

View Full Product Details