Product Details

SNP ID
rs111981667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:31993220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCAAAAGCAATGTTTTTATTGA[A/C]TATAAAAGTGTATGTTTTCAGCTGT
Phenotype
MIM: 606245
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC105371727 PubMed Links

Gene Details

Gene
LOC105371727
Gene Name
uncharacterized LOC105371727
There are no transcripts associated with this gene.

Gene
SUZ12
Gene Name
SUZ12 polycomb repressive complex 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321207.1 Intron NP_001308136.1
NM_015355.3 Intron NP_056170.2
XM_006721794.3 Intron XP_006721857.1
XM_017024409.1 Intron XP_016879898.1

View Full Product Details