Product Details

SNP ID
rs118043797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:32018522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGCAGCTGTGTGTGATAGCTTG[A/G]AAGGTTTACTGCTGCCTCAAGTCCT
Phenotype
MIM: 610432
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF125 PubMed Links

Gene Details

Gene
RNF125
Gene Name
ring finger protein 125
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017831.3 310 UTR 5 NP_060301.2
XM_011526045.2 310 UTR 5 XP_011524347.1
XM_011526046.2 310 Intron XP_011524348.1
XM_011526047.2 310 UTR 5 XP_011524349.1

View Full Product Details