Product Details

SNP ID
rs114609667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37554454 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGCTAGCTTGCATAGATTCTCA[A/C]AAGGAGTTTATGAATCTTGTTACTT
Phenotype
MIM: 613903
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF540 PubMed Links

Gene Details

Gene
ZNF540
Gene Name
zinc finger protein 540
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172225.2 Intron NP_001165696.1
NM_001172226.2 Intron NP_001165697.1
NM_152606.4 Intron NP_689819.1
Gene
ZNF571
Gene Name
zinc finger protein 571
There are no transcripts associated with this gene.

Gene
ZNF571-AS1
Gene Name
ZNF571 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details