Product Details

SNP ID
rs117156037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48384252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCTTTGGTGATATAGAGGTAGAA[A/G]AAATCGCAGTAGAGGACTGTCTGGA
Phenotype
MIM: 131235 MIM: 608373
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KDELR1 PubMed Links

Gene Details

Gene
KDELR1
Gene Name
KDEL endoplasmic reticulum protein retention receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006801.2 777 Silent Mutation TTC,TTT F194F NP_006792.1
Gene
SYNGR4
Gene Name
synaptogyrin 4
There are no transcripts associated with this gene.

View Full Product Details