Product Details

SNP ID
rs116280380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10013577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGAACTTGCAGTGCAACTGGCCC[A/G]TGACCCCAAGAAGCGCTACCAGGGT
Phenotype
MIM: 120216 MIM: 608575
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COL5A3 PubMed Links

Gene Details

Gene
COL5A3
Gene Name
collagen type V alpha 3
There are no transcripts associated with this gene.

Gene
RDH8
Gene Name
retinol dehydrogenase 8 (all-trans)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015725.2 329 Missense Mutation CAT,CGT H47R NP_056540.2

View Full Product Details