Product Details

SNP ID
rs111251683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:23376528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGCACAGAACACACCACCTCAA[C/T]GTGGGAAAGTGTATAAACATGAAAA
Phenotype
MIM: 613842 MIM: 611270
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GZF1 PubMed Links

Gene Details

Gene
GZF1
Gene Name
GDNF inducible zinc finger protein 1
There are no transcripts associated with this gene.

Gene
NAPB
Gene Name
NSF attachment protein beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283018.1 1874 UTR 3 NP_001269947.1
NM_001283020.1 1874 UTR 3 NP_001269949.1
NM_001283026.1 1874 UTR 3 NP_001269955.1
NM_022080.2 1874 UTR 3 NP_071363.1
XM_011529313.1 1874 UTR 3 XP_011527615.1
XM_011529315.2 1874 Intron XP_011527617.1
XM_017028008.1 1874 UTR 3 XP_016883497.1

View Full Product Details