Product Details

SNP ID
rs112419924
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:106941541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATCCCATCCACTTGTTCTGAGT[A/C]ATACCCGATCTCACGAAGCTCCAAG
Phenotype
MIM: 300970
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MORC4 PubMed Links

Gene Details

Gene
MORC4
Gene Name
MORC family CW-type zinc finger 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085354.2 2989 UTR 3 NP_001078823.1
NM_024657.4 2989 Missense Mutation GAC,TAC D918Y NP_078933.3
XM_005262190.3 2989 Missense Mutation GAC,TAC D826Y XP_005262247.1
XM_006724691.2 2989 Missense Mutation GAC,TAC D796Y XP_006724754.1
XM_011531027.2 2989 Missense Mutation GAC,TAC D741Y XP_011529329.1
XM_017029844.1 2989 UTR 3 XP_016885333.1

View Full Product Details