Product Details

SNP ID
rs114122128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96840473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCAGCTTGTTCACCAGCTGGCT[A/G]TGACCCTTCAGACAGGCCCAGTGCA
Phenotype
MIM: 610736 MIM: 607804
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD23 PubMed Links

Gene Details

Gene
ANKRD23
Gene Name
ankyrin repeat domain 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144994.7 497 Silent Mutation CAC,CAT H156H NP_659431.5
Gene
ANKRD39
Gene Name
ankyrin repeat domain 39
There are no transcripts associated with this gene.

Gene
CNNM3
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 3
There are no transcripts associated with this gene.

View Full Product Details