Product Details

SNP ID
rs116810459
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:88075234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGTGAGCTGTATTGCTAAATAT[C/T]GCTCAATAATTCTTAAATCCAGAAG
Phenotype
MIM: 606846
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4780 PubMed Links

Gene Details

Gene
MIR4780
Gene Name
microRNA 4780
There are no transcripts associated with this gene.

Gene
SMYD1
Gene Name
SET and MYND domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198274.3 Intron NP_938015.1
XM_005264156.2 Intron XP_005264213.1

View Full Product Details