Product Details
- SNP ID
-
rs116456722
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:32822733 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAAAAGCATAATACAAATTTTCCC[A/C]CTTACACTGCATGTCGAAGTAATTC
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC105377021
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11438895] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC105377021
- Gene Name
- uncharacterized LOC105377021
There are no transcripts associated with this gene.
- Gene
- TRIM71
- Gene Name
- tripartite motif containing 71
View Full Product Details