Product Details

SNP ID
rs114882092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44979046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGCACAGTTGTTGGTAAATGCT[C/T]AGCTCTGTGTATCACAGAGCCATGC
Phenotype
MIM: 606488
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOSC7 PubMed Links

Gene Details

Gene
EXOSC7
Gene Name
exosome component 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015004.3 Intron NP_055819.2
XM_017005930.1 Intron XP_016861419.1
Gene
ZDHHC3
Gene Name
zinc finger DHHC-type containing 3
There are no transcripts associated with this gene.

View Full Product Details