Product Details
- SNP ID
-
rs113926971
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:132490786 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCGTCCCCCGCGCCGCCGCCGCCG[A/C]GAGTCCAGCTCGCAGGAGGAGGAGG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FBRSL1
PubMed Links
Gene Details
- Gene
- FBRSL1
- Gene Name
- fibrosin like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001142641.1 |
322 |
Silent Mutation |
CGA,CGC |
R72R |
NP_001136113.1 |
XM_005266171.3 |
322 |
Silent Mutation |
CGA,CGC |
R72R |
XP_005266228.1 |
XM_005266173.3 |
322 |
Silent Mutation |
CGA,CGC |
R72R |
XP_005266230.1 |
XM_005266175.3 |
322 |
Silent Mutation |
CGA,CGC |
R72R |
XP_005266232.1 |
XM_005266176.3 |
322 |
Silent Mutation |
CGA,CGC |
R72R |
XP_005266233.1 |
XM_005266177.3 |
322 |
Silent Mutation |
CGA,CGC |
R72R |
XP_005266234.1 |
XM_005266181.3 |
322 |
Silent Mutation |
CGA,CGC |
R72R |
XP_005266238.1 |
XM_011534803.2 |
322 |
Silent Mutation |
CGA,CGC |
R72R |
XP_011533105.1 |
XM_011534804.2 |
322 |
Silent Mutation |
CGA,CGC |
R72R |
XP_011533106.1 |
XM_011534805.2 |
322 |
Silent Mutation |
CGA,CGC |
R72R |
XP_011533107.1 |
XM_011534806.2 |
322 |
Silent Mutation |
CGA,CGC |
R72R |
XP_011533108.1 |
XM_011534807.2 |
322 |
Silent Mutation |
CGA,CGC |
R72R |
XP_011533109.1 |
XM_011534808.2 |
322 |
Silent Mutation |
CGA,CGC |
R72R |
XP_011533110.1 |
XM_011534809.2 |
322 |
Silent Mutation |
CGA,CGC |
R72R |
XP_011533111.1 |
XM_011534810.2 |
322 |
Silent Mutation |
CGA,CGC |
R72R |
XP_011533112.1 |
XM_011534812.2 |
322 |
UTR 5 |
|
|
XP_011533114.1 |
XM_011534816.2 |
322 |
Silent Mutation |
CGA,CGC |
R72R |
XP_011533118.1 |
View Full Product Details