Product Details

SNP ID
rs117238776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:75989314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGCCAGTGAGGCAGACTGGTGAC[A/G]TAAGGTTGGATTACACAATTGGACA
Phenotype
MIM: 614068 MIM: 190230
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFT43 PubMed Links

Gene Details

Gene
IFT43
Gene Name
intraflagellar transport 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102564.1 Intron NP_001096034.1
NM_001255995.1 Intron NP_001242924.1
NM_052873.2 Intron NP_443105.2
Gene
TGFB3
Gene Name
transforming growth factor beta 3
There are no transcripts associated with this gene.

View Full Product Details