Product Details

SNP ID
rs117056593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:51094314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCATCTATGTGTTCATTTCTTG[C/T]AGGTCTTGATTTGTTCATCATTATT
Phenotype
MIM: 189889
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TFCP2 PubMed Links

Gene Details

Gene
TFCP2
Gene Name
transcription factor CP2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173452.1 3157 UTR 3 NP_001166923.1
NM_001173453.1 3157 UTR 3 NP_001166924.1
NM_005653.4 3157 UTR 3 NP_005644.2

View Full Product Details