Product Details

SNP ID
rs149476310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:69186543 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTGTTTTGAATGATATTTGCTT[C/T]GGTGCAGGCAAGTGTTTAGAGACTT
Phenotype
MIM: 605122
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SUPV3L1 PubMed Links

Gene Details

Gene
SUPV3L1
Gene Name
Suv3 like RNA helicase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301683.1 586 Silent Mutation TTC,TTT F19F NP_001288612.1
NM_001323584.1 586 Silent Mutation TTC,TTT F19F NP_001310513.1
NM_001323585.1 586 Missense Mutation TCG,TTG S5L NP_001310514.1
NM_001323586.1 586 Missense Mutation TCG,TTG S5L NP_001310515.1
NM_001323587.1 586 UTR 5 NP_001310516.1
NM_001323588.1 586 UTR 5 NP_001310517.1
NM_003171.4 586 Silent Mutation TTC,TTT F150F NP_003162.2

View Full Product Details