Product Details

SNP ID
rs138068705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118978820 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCGAATTGTTAAGCCACCAAAA[C/T]TACCCCTAGAGAAAAAACCCAACCC
Phenotype
MIM: 615755
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOXR1 PubMed Links

Gene Details

Gene
FOXR1
Gene Name
forkhead box R1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181721.2 325 Silent Mutation CTA,TTA L34L NP_859072.1
XM_017017575.1 325 Silent Mutation CTA,TTA L34L XP_016873064.1
XM_017017576.1 325 Silent Mutation CTA,TTA L34L XP_016873065.1

View Full Product Details