Product Details

SNP ID
rs138446220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5454001 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAGAGGCTGCCTATCTGCAGATC[C/G]AATGTTCTTTCTCACTCCTACTGCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR51B5 PubMed Links

Gene Details

Gene
OR51B5
Gene Name
olfactory receptor family 51 subfamily B member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005567.2 594 Intron NP_001005567.2
Gene
OR51I2
Gene Name
olfactory receptor family 51 subfamily I member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004754.2 594 Silent Mutation TCC,TCG S171S NP_001004754.1

View Full Product Details