Product Details

SNP ID
rs141713257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67633011 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTGTGAAGATGAAGGACTTGAA[A/G]TTCTCCTGGGCATAGCGCTCACTGT
Phenotype
MIM: 614413 MIM: 604648
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACY3 PubMed Links

Gene Details

Gene
ACY3
Gene Name
aminoacylase 3
There are no transcripts associated with this gene.

Gene
NUDT8
Gene Name
nudix hydrolase 8
There are no transcripts associated with this gene.

Gene
TBX10
Gene Name
T-box 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005995.4 1020 Silent Mutation AAC,AAT N214N NP_005986.2
XM_011544992.2 1020 Silent Mutation AAC,AAT N259N XP_011543294.1
XM_011544993.1 1020 Silent Mutation AAC,AAT N53N XP_011543295.1

View Full Product Details