Product Details

SNP ID
rs143984556
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60699591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAACCAAAACAGGAAATAAATAC[G/T]AATGAAACTGAGCTCTAAGCAGCAT
Phenotype
MIM: 606549
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
MS4A8 PubMed Links

Gene Details

Gene
MS4A8
Gene Name
membrane spanning 4-domains A8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031457.1 18 UTR 5 NP_113645.1

View Full Product Details