Product Details

SNP ID
rs145210367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64559194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTAAGCCAGTCCATCTTCATGTC[C/T]GGGATCCTGGTGGGCTCCTTTATCT
Phenotype
MIM: 607097
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC22A11 PubMed Links

Gene Details

Gene
SLC22A11
Gene Name
solute carrier family 22 member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307985.1 852 Silent Mutation TCC,TCT S151S NP_001294914.1
NM_018484.3 852 Silent Mutation TCC,TCT S151S NP_060954.1
XM_011545167.1 852 Intron XP_011543469.1

View Full Product Details