Product Details

SNP ID
rs145283432
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:7960214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAAAAAAGTCCTGGAAGCTGAT[A/G]TGGCGGAAGCTGTAGAACTTCTTGA
Phenotype
MIM: 609662
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NLRP10 PubMed Links

Gene Details

Gene
NLRP10
Gene Name
NLR family pyrin domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176821.3 1625 Silent Mutation CAC,CAT H466H NP_789791.1
XM_011520043.2 1625 Silent Mutation CAC,CAT H466H XP_011518345.1

View Full Product Details