Product Details

SNP ID
rs145740158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:106077891 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGCCCGGGACGCTAAGGCAGCC[A/T]TGGTACTACAGGTCTTTTTTGGTAT
Phenotype
MIM: 607756
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AASDHPPT PubMed Links

Gene Details

Gene
AASDHPPT
Gene Name
aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015423.2 327 Missense Mutation ATG,TTG M61L NP_056238.2
Gene
KBTBD3
Gene Name
kelch repeat and BTB domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152433.3 327 Intron NP_689646.2
NM_198439.2 327 Intron NP_940841.1
XM_006718767.3 327 Intron XP_006718830.1
XM_011542618.2 327 Intron XP_011540920.1
XM_011542619.2 327 Intron XP_011540921.1

View Full Product Details