Product Details

SNP ID
rs147661207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:106079654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTGCTTGCTGCTGAACCTGAGC[C/T]GCAAGTTGGAATTGATATAATGAAG
Phenotype
MIM: 607756
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AASDHPPT PubMed Links

Gene Details

Gene
AASDHPPT
Gene Name
aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015423.2 517 Missense Mutation CCG,CTG P124L NP_056238.2
Gene
KBTBD3
Gene Name
kelch repeat and BTB domain containing 3
There are no transcripts associated with this gene.

View Full Product Details