Product Details

SNP ID
rs149595264
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:130449253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCCCCCTCCAGGGGCTCACCGGG[G/T]GCTCTTCAGACCTGCGACGCTGCTT
Phenotype
MIM: 607509
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADAMTS15 PubMed Links

Gene Details

Gene
ADAMTS15
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139055.3 280 Missense Mutation GGC,TGC G94C NP_620686.1
XM_005271419.4 280 Intron XP_005271476.1
Gene
LOC646383
Gene Name
uncharacterized LOC646383
There are no transcripts associated with this gene.

View Full Product Details