Product Details

SNP ID
rs150608349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58214785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTTCTTCAGATCGGCAGAAATA[C/T]GCATCAAGGAAACCAAACCACCATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR1S1 PubMed Links

Gene Details

Gene
OR1S1
Gene Name
olfactory receptor family 1 subfamily S member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004458.1 Intron NP_001004458.1

View Full Product Details