Product Details

SNP ID
rs150648332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68041805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACGCTTTGTGGTTGATGTTCGGC[A/G]CTGTGAGGAGCTGGAGAAGACCTTC
Phenotype
MIM: 602141 MIM: 604592
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR4691 PubMed Links

Gene Details

Gene
MIR4691
Gene Name
microRNA 4691
There are no transcripts associated with this gene.

Gene
MIR6753
Gene Name
microRNA 6753
There are no transcripts associated with this gene.

Gene
MIR7113
Gene Name
microRNA 7113
There are no transcripts associated with this gene.

Gene
NDUFS8
Gene Name
NADH:ubiquinone oxidoreductase core subunit S8
There are no transcripts associated with this gene.

Gene
TCIRG1
Gene Name
T-cell immune regulator 1, ATPase H+ transporting V0 subunit a3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006019.3 294 Missense Mutation CAC,CGC H57R NP_006010.2
NM_006053.3 294 Intron NP_006044.1
XM_005273709.3 294 Missense Mutation CAC,CGC H57R XP_005273766.1
XM_011544726.2 294 Missense Mutation CAC,CGC H57R XP_011543028.1
XM_017017089.1 294 Missense Mutation CAC,CGC H57R XP_016872578.1
XM_017017090.1 294 UTR 5 XP_016872579.1

View Full Product Details