Product Details

SNP ID
rs139095356
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:117041196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAGGAGGGCAGAGTGGGCGGCC[A/G]CTTTACGAAAGTGTCATTTTAGGAC
Phenotype
MIM: 609073 MIM: 611585
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
FBXW8 PubMed Links

Gene Details

Gene
FBXW8
Gene Name
F-box and WD repeat domain containing 8
There are no transcripts associated with this gene.

Gene
TESC
Gene Name
tescalcin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168325.1 Intron NP_001161797.1
NM_017899.3 Intron NP_060369.3
XM_011538503.1 Intron XP_011536805.1
XM_017019532.1 Intron XP_016875021.1
XM_017019533.1 Intron XP_016875022.1

View Full Product Details