Product Details

SNP ID
rs139841107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110043423 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTGTGTGGAGAATTGTTTGACT[A/G]TAGGGGAGCAAGAGAGCAGGATGCC
Phenotype
MIM: 615123
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD13A PubMed Links

Gene Details

Gene
ANKRD13A
Gene Name
ankyrin repeat domain 13A
There are no transcripts associated with this gene.

Gene
C12orf76
Gene Name
chromosome 12 open reading frame 76
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207435.2 Intron NP_997318.1
XM_017019297.1 Intron XP_016874786.1

View Full Product Details