Product Details

SNP ID
rs142955817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:42313182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGGTTTCCATTTTTTTTGTTCT[C/T]CTTCAATTTTGGCTTGCTGTGATTC
Phenotype
MIM: 610750
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZCRB1 PubMed Links

Gene Details

Gene
ZCRB1
Gene Name
zinc finger CCHC-type and RNA binding motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033114.3 639 Missense Mutation GAA,GGA E180G NP_149105.3
XM_017020124.1 639 Missense Mutation GAA,GGA E139G XP_016875613.1

View Full Product Details