Product Details

SNP ID
rs144153566
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:102730310 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATAAGAGTTGGCTTCCAGAAAC[A/G]CACATTCCTCACTCTCACTTACTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC168 PubMed Links

Gene Details

Gene
CCDC168
Gene Name
coiled-coil domain containing 168
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146197.1 20527 Missense Mutation GCG,GTG A6796V NP_001139669.1
XM_011521106.1 20527 Missense Mutation GCG,GTG A6756V XP_011519408.1

View Full Product Details