Product Details

SNP ID
rs138055873
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43276995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCCCACCTCAGCCCTCTCAGACA[C/T]CTGTGTGGAGAGAAGTCAGCAATCC
Phenotype
MIM: 606776
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TGM7 PubMed Links

Gene Details

Gene
TGM7
Gene Name
transglutaminase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052955.2 1843 Missense Mutation ATG,GTG M614V NP_443187.1
XM_017021903.1 1843 Missense Mutation ATG,GTG M615V XP_016877392.1

View Full Product Details