Product Details

SNP ID
rs140070100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43279148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACAAGTGGGGAGGCTCCAGACAG[A/G]TATCTTTTAGGACCAGCATGGACCT
Phenotype
MIM: 606776
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TGM7 PubMed Links

Gene Details

Gene
TGM7
Gene Name
transglutaminase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052955.2 1811 Missense Mutation ACC,ATC T603I NP_443187.1
XM_017021903.1 1811 Missense Mutation ACC,ATC T604I XP_016877392.1

View Full Product Details